Assay Details
Gene Symbol: LOC4327164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795005.2 | XP_015650491.1 | 5 - 6 | 1222 | 150 | |
GenBank mRNA | AK101948.1 | - | 5 - 6 | 1223 | 150 | |
AK106555.1 | - | 5 - 6 | 1045 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
