Assay Details
Gene Symbol: LOC4327164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795005.2 | XP_015650491.1 | 6 - 7 | 1328 | 95 | |
GenBank mRNA | AK101948.1 | - | 6 - 7 | 1329 | 95 | |
AK106555.1 | - | 6 - 7 | 1151 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
