Assay Details
| Gene Symbol: LOC4327680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766670.2 | XP_015622156.1 | 1 - 2 | 267 | 68 | |
| XM_015766671.2 | XP_015622157.1 | 1 - 2 | 267 | 68 | ||
| GenBank mRNA | AJ495797.1 | - | 1 - 2 | 251 | 68 | |
More Information
Set Membership: |
|

LOC4327680
Genomic Map