Assay Details
| Gene Symbol: LOC4327201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759097.2 | XP_015614583.1 | 1 - 2 | 589 | 80 | |
| XM_015759104.2 | XP_015614590.1 | 1 - 2 | 589 | 80 | ||
| GenBank mRNA | AK242004.1 | - | 1 - 2 | 481 | 80 | |
More Information
Set Membership: |
|

LOC4327201
Genomic Map