Assay Details
| Gene Symbol: LOC4327201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759097.2 | XP_015614583.1 | 11 - 12 | 1819 | 95 | |
| XM_015759104.2 | XP_015614590.1 | 9 - 10 | 1648 | 95 | ||
More Information
Set Membership: |
|

LOC4327201
Genomic Map