Assay Details
| Gene Symbol: LOC4325550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767035.2 | XP_015622521.1 | 2 - 3 | 633 | 101 | |
| GenBank mRNA | AK071920.1 | - | 2 - 3 | 569 | 101 | |
| AK104674.1 | - | 2 - 3 | 556 | 101 | ||
More Information
Set Membership: |
|

LOC4325550
Genomic Map