Assay Details
Gene Symbol: LOC4327014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779017.2 | XP_015634503.1 | 2 - 3 | 817 | 94 | |
GenBank mRNA | AK072330.1 | - | 2 - 3 | 775 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
