Assay Details
| Gene Symbol: LOC4327899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756515.2 | XP_015612001.1 | 3 - 4 | 452 | 90 | |
| XM_015756521.2 | XP_015612007.1 | 3 - 4 | 452 | 90 | ||
| GenBank mRNA | AB179769.1 | - | 3 - 4 | 437 | 90 | |
More Information
Set Membership: |
|

LOC4327899
Genomic Map