Assay Details
| Gene Symbol: LOC4325862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765655.2 | XP_015621141.1 | 2 | 3200 | 77 | |
| GenBank mRNA | AK120325.1 | - | 1 | 3158 | 77 | |
More Information
Set Membership: |
|

LOC4325862
Genomic Map