Assay Details
| Gene Symbol: LOC4327547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765776.2 | XP_015621262.1 | 5 - 6 | 919 | 88 | |
| XM_015765785.2 | XP_015621271.1 | 5 - 6 | 916 | 88 | ||
| GenBank mRNA | AK111939.1 | - | 2 - 3 | 248 | 88 | |
More Information
Set Membership: |
|

LOC4327547
Genomic Map