Assay Details
| Gene Symbol: LOC4327493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757505.2 | XP_015612991.1 | 8 - 9 | 1505 | 74 | |
| GenBank mRNA | AK060836.1 | - | 7 - 8 | 1355 | 74 | |
| AK065059.1 | - | 8 - 9 | 1443 | 74 | ||
| KC140120.1 | - | 8 - 9 | 1434 | 74 | ||
More Information
Set Membership: |
|

LOC4327493
Genomic Map