Assay Details
Gene Symbol: LOC4327615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766601.2 | XP_015622087.1 | 2 - 3 | 931 | 101 | |
XM_015766602.2 | XP_015622088.1 | 2 - 3 | 913 | 101 | ||
GenBank mRNA | AK103249.1 | - | 2 - 3 | 854 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
