Assay Details
| Gene Symbol: LOC4327586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766947.2 | XP_015622433.1 | 8 - 9 | 1057 | 126 | |
| GenBank mRNA | AK070492.1 | - | 8 - 9 | 1031 | 126 | |
More Information
Set Membership: |
|

LOC4327586
Genomic Map