Assay Details
| Gene Symbol: LOC4324891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778363.2 | XP_015633849.1 | 9 - 10 | 1348 | 68 | |
| XM_015778432.2 | XP_015633918.1 | 10 - 11 | 763 | 68 | ||
| GenBank mRNA | AK070098.1 | - | 9 - 10 | 942 | 68 | |
More Information
Set Membership: |
|

LOC4324891
Genomic Map