Assay Details
| Gene Symbol: LOC4324361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787549.2 | XP_015643035.1 | 4 - 5 | 1063 | 65 | |
| GenBank mRNA | AK072446.1 | - | 1 - 2 | 2028 | 65 | |
More Information
Set Membership: |
|

LOC4324361
Genomic Map