Assay Details
| Gene Symbol: LOC4326952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773873.2 | XP_015629359.1 | 10 - 11 | 1335 | 81 | |
| GenBank mRNA | AK106700.1 | - | 10 - 11 | 1291 | 81 | |
More Information
Set Membership: |
|

LOC4326952
Genomic Map