Assay Details
| Gene Symbol: LOC4326952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773873.2 | XP_015629359.1 | 8 - 9 | 1200 | 105 | |
| GenBank mRNA | AK106700.1 | - | 8 - 9 | 1156 | 105 | |
More Information
Set Membership: |
|

LOC4326952
Genomic Map