Assay Details
| Gene Symbol: LOC4325394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794411.2 | XP_015649897.1 | 3 - 4 | 458 | 91 | |
| XM_015794417.2 | XP_015649903.1 | 3 - 4 | 458 | 91 | ||
| GenBank mRNA | AK070420.1 | - | 3 - 4 | 464 | 91 | |
More Information
Set Membership: |
|

LOC4325394
Genomic Map