Assay Details
| Gene Symbol: LOC4325394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794411.2 | XP_015649897.1 | 8 - 9 | 1058 | 127 | |
| XM_015794417.2 | XP_015649903.1 | 8 - 9 | 1058 | 127 | ||
| GenBank mRNA | AK070420.1 | - | 8 - 9 | 1064 | 127 | |
More Information
Set Membership: |
|

LOC4325394
Genomic Map