Assay Details
| Gene Symbol: LOC4325237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767030.2 | XP_015622516.1 | 8 - 9 | 730 | 112 | |
| GenBank mRNA | AK318565.1 | - | 1 - 2 | 89 | 112 | |
More Information
Set Membership: |
|

LOC4325237
Genomic Map