Assay Details
| Gene Symbol: LOC4326400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759471.2 | XP_015614957.1 | 3 - 4 | 589 | 98 | |
| GenBank mRNA | AK102519.1 | - | 1 - 2 | 35 | 98 | |
More Information
Set Membership: |
|

LOC4326400
Genomic Map