Assay Details
| Gene Symbol: LOC4326400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759461.2 | XP_015614947.1 | 8 - 9 | 1786 | 138 | |
| XM_015759471.2 | XP_015614957.1 | 8 - 9 | 1777 | 138 | ||
| GenBank mRNA | AK102519.1 | - | 6 - 7 | 1223 | 138 | |
More Information
Set Membership: |
|

LOC4326400
Genomic Map