Assay Details
| Gene Symbol: LOC4326400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759461.2 | XP_015614947.1 | 9 - 10 | 2754 | 90 | |
| XM_015759471.2 | XP_015614957.1 | 9 - 10 | 2745 | 90 | ||
| GenBank mRNA | AK102519.1 | - | 7 - 8 | 2191 | 90 | |
More Information
Set Membership: |
|

LOC4326400
Genomic Map