Assay Details
| Gene Symbol: LOC4327984 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771863.2 | XP_015627349.1 | 19 - 20 | 2353 | 87 | |
| GenBank mRNA | AK102833.1 | - | 19 - 20 | 2279 | 87 | |
More Information
Set Membership: |
|

LOC4327984
Genomic Map