Assay Details
| Gene Symbol: LOC9269307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768171.2 | XP_015623657.1 | 17 - 18 | 3130 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9269307| Gene Symbol: LOC9269307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768171.2 | XP_015623657.1 | 17 - 18 | 3130 | 150 | |
Set Membership: |
|