Assay Details
| Gene Symbol: LOC4328251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771546.2 | XP_015627032.1 | 13 - 14 | 1336 | 87 | |
| GenBank mRNA | AK073412.1 | - | 13 - 14 | 1287 | 87 | |
| AK103804.1 | - | 13 - 14 | 1288 | 87 | ||
More Information
Set Membership: |
|

LOC4328251
Genomic Map