Assay Details
| Gene Symbol: LOC4328251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771546.2 | XP_015627032.1 | 4 - 5 | 683 | 71 | |
| GenBank mRNA | AK073412.1 | - | 4 - 5 | 634 | 71 | |
| AK103804.1 | - | 4 - 5 | 635 | 71 | ||
More Information
Set Membership: |
|

LOC4328251
Genomic Map