Assay Details
| Gene Symbol: LOC4328286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770974.1 | XP_015626460.1 | 1 - 2 | 290 | 90 | |
| GenBank mRNA | AK073620.1 | - | 1 - 2 | 188 | 90 | |
| KC140119.1 | - | 1 - 2 | 295 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770974.1 transcripts.
Set Membership: |
|

LOC4328286
Genomic Map