Assay Details
| Gene Symbol: LOC4328405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768309.2 | XP_015623795.1 | 7 - 8 | 726 | 69 | |
| GenBank mRNA | AK062817.1 | - | 2 - 3 | 140 | 69 | |
More Information
Set Membership: |
|

LOC4328405
Genomic Map