Assay Details
| Gene Symbol: LOC4328405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768309.2 | XP_015623795.1 | 8 - 9 | 796 | 60 | |
| GenBank mRNA | AK062817.1 | - | 3 - 4 | 210 | 60 | |
More Information
Set Membership: |
|

LOC4328405
Genomic Map