Assay Details
| Gene Symbol: LOC4328787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771884.2 | XP_015627370.1 | 4 - 5 | 967 | 131 | |
| GenBank mRNA | AK288680.1 | - | 4 - 5 | 929 | 131 | |
More Information
Set Membership: |
|

LOC4328787
Genomic Map