Assay Details
| Gene Symbol: LOC4346937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755141.2 | XP_015610627.1 | 2 - 3 | 709 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4346937| Gene Symbol: LOC4346937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755141.2 | XP_015610627.1 | 2 - 3 | 709 | 77 | |
Set Membership: |
|