Assay Details
| Gene Symbol: LOC4346967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795666.2 | XP_015651152.1 | 4 - 5 | 973 | 120 | |
| GenBank mRNA | AK120537.1 | - | 4 - 5 | 917 | 120 | |
More Information
Set Membership: |
|

LOC4346967
Genomic Map