Assay Details
Gene Symbol: LOC4346967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795666.2 | XP_015651152.1 | 5 - 6 | 1056 | 111 | |
GenBank mRNA | AK120537.1 | - | 5 - 6 | 1000 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
