Assay Details
| Gene Symbol: LOC4347182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756136.1 | XP_015611622.1 | 3 - 4 | 1322 | 102 | |
| GenBank mRNA | AB095097.1 | - | 2 - 3 | 1384 | 102 | |
| AK100279.1 | - | 3 - 4 | 1100 | 102 | ||
More Information
Set Membership: |
|

LOC4347182
Genomic Map