Assay Details
| Gene Symbol: LOC4347182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756136.1 | XP_015611622.1 | 5 - 6 | 1837 | 105 | |
| GenBank mRNA | AB095097.1 | - | 4 - 5 | 1899 | 105 | |
| AK100279.1 | - | 5 - 6 | 1615 | 105 | ||
More Information
Set Membership: |
|

LOC4347182
Genomic Map