Assay Details
| Gene Symbol: LOC4348062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759416.2 | XP_015614902.1 | 2 - 3 | 3067 | 71 | |
| XM_015759417.2 | XP_015614903.1 | 2 - 3 | 3006 | 71 | ||
| GenBank mRNA | AK073424.1 | - | 2 - 3 | 3023 | 71 | |
More Information
Set Membership: |
|

LOC4348062
Genomic Map