Assay Details
| Gene Symbol: LOC4348305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759391.2 | XP_015614877.1 | 6 - 7 | 777 | 70 | |
| GenBank mRNA | AK072694.1 | - | 6 - 7 | 725 | 70 | |
| KC140130.1 | - | 6 - 7 | 747 | 70 | ||
More Information
Set Membership: |
|

LOC4348305
Genomic Map