Assay Details
| Gene Symbol: LOC4348305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759391.2 | XP_015614877.1 | 7 - 8 | 897 | 146 | |
| GenBank mRNA | AK072694.1 | - | 7 - 8 | 845 | 146 | |
| KC140130.1 | - | 7 - 8 | 867 | 146 | ||
More Information
Set Membership: |
|

LOC4348305
Genomic Map