Assay Details
| Gene Symbol: LOC4348305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759391.2 | XP_015614877.1 | 8 - 9 | 1026 | 86 | |
| GenBank mRNA | AK072694.1 | - | 8 - 9 | 974 | 86 | |
| KC140130.1 | - | 8 - 9 | 996 | 86 | ||
More Information
Set Membership: |
|

LOC4348305
Genomic Map