Assay Details
| Gene Symbol: LOC4348337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758316.2 | XP_015613802.1 | 11 - 12 | 1348 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4348337| Gene Symbol: LOC4348337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758316.2 | XP_015613802.1 | 11 - 12 | 1348 | 105 | |
Set Membership: |
|