Assay Details
| Gene Symbol: LOC4348337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758316.2 | XP_015613802.1 | 5 - 6 | 629 | 59 | |
| GenBank mRNA | AK119350.1 | - | 5 - 6 | 635 | 59 | |
More Information
Set Membership: |
|

LOC4348337
Genomic Map