Assay Details
| Gene Symbol: LOC4349333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757426.2 | XP_015612912.1 | 1 - 2 | 157 | 92 | |
| XM_015757428.2 | XP_015612914.1 | 1 - 2 | 157 | 92 | ||
| GenBank mRNA | AB277360.1 | - | 1 - 2 | 116 | 92 | |
| AK069361.1 | - | 1 - 2 | 121 | 92 | ||
| AY570728.1 | - | 78 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757426.2, XM_015757428.2 transcripts.
Set Membership: |
|

LOC4349333
Genomic Map