Assay Details
| Gene Symbol: LOC4349333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757426.2 | XP_015612912.1 | 4 - 5 | 883 | 74 | |
| XM_015757428.2 | XP_015612914.1 | 4 - 5 | 898 | 74 | ||
| XM_015757429.2 | XP_015612915.1 | 3 - 4 | 733 | 74 | ||
| GenBank mRNA | AB277360.1 | - | 4 - 5 | 842 | 74 | |
| AK069361.1 | - | 4 - 5 | 847 | 74 | ||
More Information
Set Membership: |
|

LOC4349333
Genomic Map