Assay Details
| Gene Symbol: LOC4349333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757426.2 | XP_015612912.1 | 5 - 6 | 946 | 119 | |
| XM_015757428.2 | XP_015612914.1 | 5 - 6 | 961 | 119 | ||
| XM_015757429.2 | XP_015612915.1 | 4 - 5 | 796 | 119 | ||
| GenBank mRNA | AB277360.1 | - | 5 - 6 | 905 | 119 | |
| AK069361.1 | - | 5 - 6 | 910 | 119 | ||
More Information
Set Membership: |
|

LOC4349333
Genomic Map