Assay Details
| Gene Symbol: LOC4349393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759401.2 | XP_015614887.1 | 4 - 5 | 2240 | 92 | |
| XM_015759402.2 | XP_015614888.1 | 3 - 4 | 3234 | 92 | ||
| XM_015759403.2 | XP_015614889.1 | 3 - 4 | 3200 | 92 | ||
| GenBank mRNA | AK064321.1 | - | 2 - 3 | 345 | 92 | |
More Information
Set Membership: |
|

LOC4349393
Genomic Map