Assay Details
| Gene Symbol: LOC4349399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758804.2 | XP_015614290.1 | 4 - 5 | 1090 | 65 | |
| GenBank mRNA | AK122086.1 | - | 4 - 5 | 1086 | 65 | |
More Information
Set Membership: |
|

LOC4349399
Genomic Map