Assay Details
| Gene Symbol: LOC9269678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764229.2 | XP_015619715.1 | 4 - 5 | 657 | 62 | |
| XM_015764230.2 | XP_015619716.1 | 2 - 3 | 319 | 62 | ||
More Information
Set Membership: |
|

LOC9269678
Genomic Map