Assay Details
| Gene Symbol: LOC4351854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762627.2 | XP_015618113.1 | 2 - 3 | 479 | 104 | |
| XM_015762628.2 | XP_015618114.1 | 2 - 3 | 477 | 104 | ||
| GenBank mRNA | AK068145.1 | - | 2 - 3 | 485 | 104 | |
More Information
Set Membership: |
|

LOC4351854
Genomic Map