Assay Details
| Gene Symbol: LOC4352411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765194.2 | XP_015620680.1 | 8 - 9 | 1435 | 90 | |
| XM_015765195.2 | XP_015620681.1 | 8 - 9 | 1426 | 90 | ||
| XM_015765197.2 | XP_015620683.1 | 8 - 9 | 1435 | 90 | ||
| XM_015765198.2 | XP_015620684.1 | 8 - 9 | 1426 | 90 | ||
More Information
Set Membership: |
|

LOC4352411
Genomic Map